Haloacetic acid mix (9 components)
Catalog # REST31896
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in methyl tert-butyl ether
- Pk:1
- Description:Haloacetic acid mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This consists of bromochloroacetic acid, bromodichloroacetic acid, chlorodibromoacetic acid, dibromoacetic acid, dichloroacetic acid, monobromoacetic acid, monochloroacetic acid, tribromoacetic acid and trichloroacetic acid.