Order Entry
Germany
ContactUsLinkComponent
Weathered diesel fuel #2 kit
Weathered diesel fuel #2 kit
Catalog # REST31239
Supplier:  Restek
CAS Number:  
undefined
Weathered diesel fuel #2 kit
Catalog # REST31239
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Weathered diesel fuel #2 kit

Specifications

About this item

The kit consists of different standards.

  • Contains 1 ml each of these mixtures,
  • Diesel fuel #2 standard
  • Diesel fuel #2 standard - 25% weathered
  • Diesel fuel #2 standard - 50% weathered
  • Diesel fuel #2 standard - 75% weathered
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.