AMPA (glyphosate metabolite)
Catalog # REST32428
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 μg/ml in DI water
- Pk:1 mL
- CAS No.:1066-51-9
- Description:Aminomethyl phosphonic acid
- Formula:CH₆NO₃P
- Description:Aminomethyl phosphonic acid (AMPA)
- Pk Info:1 ml/ampoule