Order Entry
Germany
ContactUsLinkComponent
602 Purgeable aromatics calibration mix (7 components)
602 Purgeable aromatics calibration mix (7 components)
Catalog # REST30035
Supplier:  Restek
CAS Number:  
undefined
602 Purgeable aromatics calibration mix (7 components)
Catalog # REST30035
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    Purgeable aromatics calibration mix, 602
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: Benzene (71-43-2), Chlorobenzene (108-90-7), 1,2-Dichlorobenzene (95-50-1), 1,3-Dichlorobenzene (541-73-1), 1,4-Dichlorobenzene (106-46-7), Ethylbenzene (100-41-4), Toluene (108-88-3).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.