PAH supplement mix for method 8100 (8 components)
Catalog # REST31857
Supplier: Restek
CAS Number:
Specifications
- Pk:1 Ampoul
- Description:PAH supplement mix for method 8100, 1000 μg/ml each in methylene chloride
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of benzo(j)fluoranthene, dibenz(a,h)acridine, dibenz(a,j)acridine, 7H-dibenzo(c,g)carbazole, dibenzo(a,e)pyrene, dibenzo(a,h)pyrene, dibenzo(a,i)pyrene, 3-methylcholanthrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.