ContactUsLinkComponent
Standard mixture stock solution (2 components)
Standard mixture stock solution (2 components)
Catalog # REST31630
Supplier:  Restek
CAS Number:  
undefined
Standard mixture stock solution (2 components)
Catalog # REST31630
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    Standard mixture stock solution, 5000 μg/ml each in cyclohexane
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
For GC analysis of total petroleum hydrocarbons (TPH) in water.
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil
The mix consists of diesel #2 (additive-free) and mineral oil (additive-free [ USP grade] bp 325 - 460 or C18 - C32 retention time range).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.