Quality control standard mixture, revised (2 components)
Catalog # REST31641
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:Quality control standard mixture, revised, 500 μg/ml each in acetone
- Pk Info:1 ml/ampoule
Specifications
About this item
Determination of hydrocarbon oil index - applicable to drinking, surface, waste, and treated water.
- Updated reference materials for GC analysis of TPH in water
- Total hydrocarbon concentration is 1000 μg/ml
The mix consists of diesel #2 (additive-free), motor oil (additive-free bp 325 - 460 or C18 - C32 retention time range).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.