ContactUsLinkComponent
CLP 04.1 phenols calibration mix (14 components)
CLP 04.1 phenols calibration mix (14 components)
Catalog # REST31494
Lieferant:  Restek
CAS-Nummer:  
undefined
CLP 04.1 phenols calibration mix (14 components)
Catalog # REST31494
Lieferant:  Restek
CAS-Nummer:  
Spezifikationen
  • Pk:
    1
  • Description:
    CLP 04.1 phenols calibration mix, 2000 μg/ml each in methylene chloride
  • Pk Info:
    1 ml/ampoule
Spezifikationen
Über diesen Artikel
The mix consists of 4-chloro-3-methylphenol, 2-chlorophenol, 2,4-dichlorophenol, 2,4-dimethylphenol, 4,6-dinitro-2-methylphenol, 2,4-dinitrophenol, 2-methylphenol, 4-methylphenol, 2-nitrophenol, 4-nitrophenol, pentachlorophenol, phenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.