Haloacetic acid mix #2 (9 components)
REST31646
: Restek
:
- Concentration:In methyl tert-butyl ether
- Pk:1 mL
- Description:Haloacetic acid mix #2
- Pk Info:1 ml/ampoule
The mix consists of, bromochloroacetic acid (400 μg/ml), bromodichloroacetic acid (400 μg/ml), chlorodibromoacetic acid (1000 μg/ml), dibromoacetic acid (200 μg/ml), dichloroacetic acid (600 μg/ml), monobromoacetic acid (400 μg/ml), monochloroacetic acid (600 μg/ml), tribromoacetic acid (2000 μg/ml), trichloroacetic acid (200 μg/ml)
- Prepared from purified neat reference materials for the highest quality
- Lot-to-lot consistency ensures analytical reproducibility
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Meets Stage 1 requirements for US EPA Methods 552, 552.1 and 552.2 and meets requirements for Stage 2 DBPR.