Haloacetic acid mix #1 (6 components)
REST31644
: Restek
:
- Concentration:2000 μg/ml each in methyl tert-butyl ether
- Pk:1 mL
- Standards:EPA method 552, 552.1, 552.2
- Description:Haloacetic acid mix #1
- Pk Info:1 ml/ampoule
The mix consists of bromochloroacetic acid, dibromoacetic acid, dichloroacetic acid, monobromoacetic acid, monochloroacetic acid, trichloroacetic acid.
- Prepared from purified neat reference materials for the highest quality
- Lot-to-lot consistency ensures analytical reproducibility
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements. Meet Stage 1 requirements for US EPA Methods 552, 552.1 and 552.2 and meet requirements for Stage 2 DBPR.