ContactUsLinkComponent
524 Rev. 4.0 VOA kit #2
  REST30447
 :  Restek
 :  
undefined
524 Rev. 4.0 VOA kit #2
  REST30447
 :  Restek
 :  
  • Concentration:
    2000 μg/ml
  • Pk:
    1 KIT
  • Description:
    524 Rev. 4.0 VOA kit #2, contains 1 ml each of mixtures
VOA kit #2 contains 502.2 calibration mix #1: bromomethane (methyl bromide), chloroethane (ethyl chloride), chloromethane (methyl chloride), dichlorodifluoromethane (CFC-12), trichlorofluoromethane (CFC-11), vinyl chloride; 502.2 MegaMix standard: benzene, bromobenzene, bromochloromethane, bromodichloromethane, bromoform, n-butylbenzene, sec-butylbenzene, tert-butylbenzene, carbon tetrachloride, chlorobenzene, chloroform, 2-chlorotoluene, 4-chlorotoluene, dibromochloromethane, 1,2-dibromo-3-chloropropane , 1,2-dibromoethane, dibromomethane, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, 1,3-dichloropropane, 2,2-dichloropropane, 1,1-dichloropropene, cis-1,3-dichloropropene, trans-1,3-dichloropropene, ethylbenzene, hexachloro-1,3-butadiene , isopropylbenzene , 4-isopropyltoluene , methylene chloride , naphthalene, n-propylbenzene, styrene, 1,1,1,2-tetrachloroethane, 1,1,2,2-tetrachloroethane, tetrachloroethene, toluene, 1,2,3-trichlorobenzene, 1,2,4-trichlorobenzene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, 1,2,3-trichloropropane, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.524 calibration mix #7A: acetone, 2-butanone (MEK), 2-hexanone, 4-methyl-2-pentanone (MIBK), tetrahydrofuran; 524 calibration mix #7B: acrylonitrile, allyl chloride (3-chloropropene), ethyl methacrylate, methyl acrylate, methyl methacrylate, nitrobenzene, pentachloroethane; calibration mix #8: carbon disulfide, chloroacetonitrile, 1-chlorobutane (butyl chloride), trans-1,4-dichloro-2-butene, 1,1-dichloro-2-propanone, diethyl ether (ethyl ether), hexachloroethane, iodomethane (methyl iodide), methacrylonitrile, methyl tert-butyl ether (MTBE), 2-nitropropane, propionitrile; internal standard/surrogate mix: 1-bromo-4-fluorobenzene (BFB), 1,2-dichlorobenzene-d4, fluorobenzene.
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.