EP 2.4.22 composition of fatty acids by GC mix 2 (2 components)
REST35101
: Restek
:
- Pk:1
- Description:EP 2.4.22 composition of fatty acids by GC mixture 2
- Pk Info:100 mg total
The composition of fatty acids by GC mix 2 consists of five components they are, (C6:0) methyl caproate 10%, (C8:0) methyl caprylate, 10%, (C10:0) methyl decanoate 20%, (C12:0) methyl dodecanoate, 20% and (C14:0) methyl myristate 40%.