Order Entry
Germany
ContactUsLinkComponent
SOM01.1 deuterated monitoring compound mix SIM compounds (2 components)
SOM01.1 deuterated monitoring compound mix SIM compounds (2 components)
Catalog # REST33913
Supplier:  Restek
CAS Number:  
undefined
SOM01.1 deuterated monitoring compound mix SIM compounds (2 components)
Catalog # REST33913
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    SOM01.1 deuterated monitoring compound mixture SIM compounds
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The deuterated monitoring compound mix consists of fluoranthene-d10 and 2-methylnaphthalene-d10.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.