Order Entry
Germany
ContactUsLinkComponent
502.2 VOA calibration kit #3
502.2 VOA calibration kit #3
Catalog # REST30446
Supplier:  Restek
CAS Number:  
undefined
502.2 VOA calibration kit #3
Catalog # REST30446
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 μg/ml
  • Pk:
    1 KIT
  • Description:
    502.2 VOA calibration kit #3, contains 1 ml each of mixtures

Specifications

About this item

502.2 VOA calibration kit #2 contains 502.2 calibration mix #1: bromomethane (methyl bromide), chloroethane (ethyl chloride), chloromethane (methyl chloride), dichlorodifluoromethane (CFC-12), trichlorofluoromethane (CFC-11), vinyl chloride; 502.2 MegaMix standard: benzene, bromobenzene, bromochloromethane, bromodichloromethane, bromoform, n-butylbenzene, sec-butylbenzene, tert-butylbenzene, carbon tetrachloride, chlorobenzene, chloroform, 2-chlorotoluene, 4-chlorotoluene, dibromochloromethane, 1,2-dibromo-3-chloropropane , 1,2-dibromoethane, dibromomethane, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, 1,3-dichloropropane, 2,2-dichloropropane, 1,1-dichloropropene, cis-1,3-dichloropropene, trans-1,3-dichloropropene, ethylbenzene, hexachloro-1,3-butadiene , isopropylbenzene , 4-isopropyltoluene , methylene chloride , naphthalene, n-propylbenzene, styrene, 1,1,1,2-tetrachloroethane, 1,1,2,2-tetrachloroethane, tetrachloroethene, toluene, 1,2,3-trichlorobenzene, 1,2,4-trichlorobenzene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, 1,2,3-trichloropropane, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.