Order Entry
Germany
Orders LinkContactUsLinkComponent
MCPA

MCPA

MCPA, 1000 μg/ml in methanol, 1 ml/ampoule, 1 mL
Catalog # REST32269
Supplier:   Restek
MCPA
Catalog # REST32269
Supplier:  Restek
CAS Number:  
MCPA
MCPA

MCPA, 1000 μg/ml in methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
54,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 μg/ml in methanol
  • Pk:
    1 mL
  • Description:
    MCPA
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32269
  • Formula:
    C₉H₉ClO₃
  • Description:
    MCPA
  • CAS No.:
    94-74-6

Specifications