Order Entry
Germany
ContactUsLinkComponent
WA EPH aromatic hydrocarbon standard (18 components)
WA EPH aromatic hydrocarbon standard (18 components)
Catalog # REST31469
Supplier:  Restek
CAS Number:  
undefined
WA EPH aromatic hydrocarbon standard (18 components)
Catalog # REST31469
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    WA EPH aromatic hydrocarbon standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(k)fluoranthene (207-08-9); Benzo(ghi)perylene (191-24-2); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); 2-Methylnaphthalene (91-57-6); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0); 1,2,3-Trimethylbenzene (526-73-8).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.