Order Entry
Germany
ContactUsLinkComponent
Herbicide lab performance check mix (5 components)
Herbicide lab performance check mix (5 components)
Catalog # REST32063
Supplier:  Restek
CAS Number:  
undefined
Herbicide lab performance check mix (5 components)
Catalog # REST32063
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In methyl tert-butyl ether
  • Pk:
    1 mL
  • Description:
    Herbicide lab performance check mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This mix contains five components.

Components are dinoseb methyl ether (4 μg/ml), DCAA methyl ester (500 μg/ml), 4,4'-dibromooctafluorobiphenyl (250 μg/ml), 3,5-dichlorobenzoic acid methyl ester (600 µg/ml), 4-nitroanisole (1600 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.