ContactUsLinkComponent
Phenol calibration mix, EPA 528
Phenol calibration mix, EPA 528
Catalog # REST31694
Supplier:  Restek
CAS Number:  
undefined
Phenol calibration mix, EPA 528
Catalog # REST31694
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 Ampoul
  • Description:
    Phenol calibration mix, EPA 528, 2000 µg/ml each
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
This phenol calibration mix contain 12 components: 4-Chloro-3-methylphenol, 2-Chlorophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-Dinitrophenol, 2-Methylphenol (o-cresol), 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, Phenol and 2,4,6-Trichlorophenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.