Order Entry
Germany
ContactUsLinkComponent
OLC 03.2 SVOA deuterated monitoring compounds (DMC) (16 components)
OLC 03.2 SVOA deuterated monitoring compounds (DMC) (16 components)
Catalog # REST31810
Supplier:  Restek
CAS Number:  
undefined
OLC 03.2 SVOA deuterated monitoring compounds (DMC) (16 components)
Catalog # REST31810
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/m each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    OLC 03.2 SVOA deuterated monitoring compounds (DMC)
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This OLC 03.2 is an analytical method for the detection of low-concentration volatile, semivolatile, and pesticide/Aroclor organics in aqueous samples using deuterated monitoring compounds (DMCs) as sample-by-sample accuracy indicators.

It contains 16 components include: Acenaphthylene-d8, Anthracene-d10, Benzo(a)pyrene-d12, Bis-(2-chloroethyl)ether-d8, 4-Chloroaniline-d4, 2-Chlorophenol-d4, 2,4-Dichlorophenol-d3, Dimethylphthalate-d6, 4,6-Dinitro-2-methylphenol-d2, Fluorene-d10, 4-Methylphenol-d8, Nitrobenzene-d5, 2-Nitrophenol-d4, 4-Nitrophenol-d4, Phenol-d5 and Pyrene-d10.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.