NJDEP EPH 10/08 rev. 2 aromatics calibration standard (18 components)
Catalog # REST30541
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Pk:1 mL
- Description:NJDEP EPH 10/08 rev. 2 aromatics calibration standard
- Pk Info:1 ml/ampoule
Specifications
About this item
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, 2-methylnaphthalene, naphthalene, phenanthrene, 1,2,3-trimethylbenzene and pyrene
18 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.