Order Entry
Germany
ContactUsLinkComponent
NJDEP EPH 10/08 Rev. 2 aliphatics fractionation check mix (18 components)
NJDEP EPH 10/08 Rev. 2 aliphatics fractionation check mix (18 components)
Catalog # REST30544
Supplier:  Restek
CAS Number:  
undefined
NJDEP EPH 10/08 Rev. 2 aliphatics fractionation check mix (18 components)
Catalog # REST30544
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    400 µg/ml each in hexane
  • Pk:
    1 Ampoul
  • Description:
    NJDEP EPH 10/08 rev. 2 aliphatics fractionation check mix
  • Pk Info:
    5 ml/ampoule

Specifications

About this item

Consists of 18 components, those are n-nonane, n-decane,  n-dodecane,  n-tetradecane, n-hexadecane, n-octadecane,  n-eicosane,  n-heneicosane,  n-docosane,  n-tetracosane,  n-hexacosane,  n-octacosane,  n-triacontane,  n-dotriacontane,  n-tetratriacontane,  n-hexatriacontane, n-octatriacontane and n-tetracontane.

18 components

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.