Benzidine
Catalog # REST31441
Supplier: Restek
CAS Number:
Specifications
- Concentration:1,00 µg/ml in methanol
- Pk:1 mL
- CAS No.:92-87-5
- Description:Benzidine
- Formula:C₁₂H₁₂N₂
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.