1,2,4-Trichlorobenzene
REST31439
: Restek
:
- Concentration:1000 µg/ml in methanol
- Pk:1 mL
- CAS No.:120-82-1
- Description:1,2,4-Trichlorobenzene
- Formula:C₆H₃Cl₃
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.