ASTM® D3606 calibration standard #5 with sec-butanol internal standard (3 components)
Catalog # REST30665
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:ASTM D3606 calibration standard #5 with sec-butanol internal standard
- Pk Info:1 ml/ampoule
Specifications
About this item
Consists of three components, those are benzene, 2-butanol and toluene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.