Order Entry
Germany
ContactUsLinkComponent
Methylated 515.4 chlorinated acids mix (16 components)
Methylated 515.4 chlorinated acids mix (16 components)
Catalog # REST32444
Supplier:  Restek
CAS Number:  
undefined
Methylated 515.4 chlorinated acids mix (16 components)
Catalog # REST32444
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In methyl tert-butyl ether
  • Pk:
    1 mL
  • Description:
    Methylated chlorinated acid mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains Acifluorfen methyl ester, Bentazon methyl ester, Chloramben methyl ester, Dalapon methyl ester, 2,4-D methyl ester, 2,4-DB methyl ester, DCPA methyl ester (Chlorthal-dimethyl), Dicamba methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Dichlorprop methyl ester, Dinoseb methyl ether, Pentachloroanisole, Picloram methyl ester, Quinclorac methyl ester, 2,4,5-T methyl ester, 2,4,5-TP.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.