Order Entry
Germany
ContactUsLinkComponent
Herbicide reference standard, 2,4-D (2,4-dichlorophenoxyacetic acid) 1000 ?g/ml in methanol
Herbicide reference standard, 2,4-D (2,4-dichlorophenoxyacetic acid) 1000 ?g/ml in methanol
  REST32239
 :  Restek
 :  
undefined
Herbicide reference standard, 2,4-D (2,4-dichlorophenoxyacetic acid) 1000 ?g/ml in methanol
  REST32239
 :  Restek
 :  

 

  • Concentration:
    1000 µg/ml in methanol
  • Pack type:
    Ampoule
  • Pk:
    1
  • CAS No.:
    94-75-7
  • Description:
    2,4-D (2,4-Dichlorophenoxyacetic acid)
  • Formula:
    C₈H₆Cl₂O₃
  • Description:
    2,4-D (2,4-Dichlorophenoxyacetic acid)
  • Pk Info:
    1 mL/ampoule

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.