Atlantic RBCA VPH mix (12 components)
Catalog # REST31871
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml in P and T methanol
- Pk:1 Ampoul
- Description:Atlantic RBCA VPH mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: n-hexane, C6, n-heptane, C7, n-octane, C8, n-decane, C10, benzene, ethylbenzene, 1-methyl-3-ethylbenzene, toluene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, o-xylene, p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.