ContactUsLinkComponent
GC multi-residue pesticide standard #8 (24 components)
GC multi-residue pesticide standard #8 (24 components)
Catalog # REST32570
Supplier:  Restek
CAS Number:  
undefined
GC multi-residue pesticide standard #8 (24 components)
Catalog # REST32570
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #8, 24 components, 100 μg/ml each in toluene
  • Pk Info:
    1 ml /ampoule
Specifications
About this item
The multiresidue accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability
The multiresidue pesticide standard consist of 24 components bromfenvinfos-methyl, bromfenvinphos, bromophos ethyl, bromophos methyl, carbophenothion, chlorfenvinphos, chlorthiophos, coumaphos, edifenphos, ethion, fenamiphos, fenchlorphos (ronnel), fenthion, iodofenphos, leptophos, malathion, methacrifos, profenofos, prothiofos, sulphotepp, sulphrofos, terbufos, tetrachlorvinfos, tolclofos-methyl.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.