GC column connectors, Defined PressFit
Supplier: Restek
Certificates
About this item
These PressFit connectors feature ID-specific ends for better sealability and a defined fit.
- Deactivated borosilicate glass
- Connect a guard column to an analytical column
- Repair a broken column
- Connect a column outlet to a transfer line
These connectors are colour coded for easy identification.