Alkaline detergents, powder, LaboClean PLM, neodisher®
Catalog # 148-3133
Supplier: Dr. Weigert
Specifications
- Description:LaboClean PLM, alkaline detergent
- Seq:02
- Pack type:Bucket
- Pk:10 kg
- pH range:11,1 - 11,6
Specifications
About this item
Mildly alkaline detergent.
- Free of oxidative additives
Suitable for the removal of the basic components of creams respective lotions and easily emulsified residues. With defoaming properties.