Order Entry
Germany
ContactUsLinkComponent
Bovine Ribonuclease A (from Pancreas)
Bovine Ribonuclease A (from Pancreas)
Catalog # 70856-3
CAS Number:  
Bovine Ribonuclease A (from Pancreas)
Catalog # 70856-3
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 mL
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    15...30 °C
  • Form:
    Solution
  • Enzyme name:
    Ribonuclease A
  • Concentration:
    10 mg/ml
  • pH:
    pH 7,5
  • Grade:
    High purity

Specifications

About this item

Convenient solution for selective degradation of RNA.

This is a convenient alternative to powdered RNase A. It is a highly purified preparation of bovine pancreatic ribonuclease A suitable for use in selective removal of RNA. It has been pretreated to remove DNase I and is suitable for RNA digestion in plasmid purification procedures.

Supplied at a concentration of 10 mg/ml in 10 mM Tris-HCl, 1 mM EDTA, 50% glycerol, pH 7,5.