Order Entry
Germany
ContactUsLinkComponent
Trichoderma viride cellulase
Trichoderma viride cellulase
Certificates
Trichoderma viride cellulase
Certificates

Specifications

  • CAS No.:
    9012-54-8
  • Species:
    Trichoderma viride
  • Storage conditions:
    2...8 °C
  • Form:
    Yellowish white solid
  • Enzyme name:
    Cellulase
  • Enzyme activity:
    ≥10000 U/g
  • pH:
    4,0

Specifications

Related Information

Frequently Bought Together