Sterilisation Bags
Supplier: MEGRO
Certificates
About this item
These bags are ideal for packing various products like clothes, trays, compresses and medical instruments which are subjected to sterilisation by steam, gas (EO and FO).
- Unsuitable for gamma, hot-air and hydrogen peroxide sterilisation
- Paper has a weight of 68 g/m²
The foil consists of a 12 µm PET and 38 µm PP laminate and has three indicators for steam, EO gas, formaldehyde-sterilisation.
Certifications: Sterilisation package according to the requirements of ISO 11607/EN 868.
Caution: Unsuitable for gamma, hot-air and hydrogen peroxide sterilisation.