Gloves, natural rubber, Sicco
Supplier: Bohlender
Certificates
About this item
These natural rubber gloves are wearable on both sides.
- Perforation and abration proof
- High flexibility
- Low voltage value
- Low electrostatic charging
Specifications
- Catalog No:
- BOHLV1972-09
- BOHLV1972-07
- BOHLV1972-08
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet