Order Entry
Germany
ContactUsLinkComponent
8140/8141 Internal standards and surrogates, FPD detector
8140/8141 Internal standards and surrogates, FPD detector
Supplier:  Restek
Certificates
undefined
8140/8141 Internal standards and surrogates, FPD detector
Supplier:  Restek

Specifications

  • CAS No.:
    115-86-6
  • Formula:
    (C₆H₅O)₃PO
  • Description:
    Triphenyl phosphate

Specifications

Frequently Bought Together