Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
PEG-200 standard
PEG-200 standard
PEG-200 standard
REST30549
:
Restek
:
PEG-200 standard
REST30549
:
Restek
:
Pk:
1 Ampoul
CAS No.:
25322-68-3
Description:
PEG-200 standard, neat
Formula:
(CH₂OHCH₂OH)n
Description:
PEG 200
Pk Info:
10 ml/ampoule