Carbamate pesticide calibration mixture
Catalog # REST32273
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:531.1 Carbamate Pesticide Calibration Mixture, 100 µg/ml each in methanol
- Pk Info:1 ml/ampoule
Specifications
About this item
US EPA 531.1 method for carbamate pesticide calibration
This calibration mixture consists 1 ml of aldicarb, aldicarb sulfone, aldicarb sulfoxide, carbaryl (Sevin), carbofuran, 3-hydroxycarbofuran, methiocarb, methomyl, oxamyl, propoxur (Baygon).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.