QuEChERS quality control standards for GC-MS analysis
Supplier: Restek
Certificates
About this item
Pesticide analysis is fast and simple using QuEChERS methods. Use these cost-effective QuEChERS standards for even greaterlab efficiency. Standards are compatible with all major methods, including mini-multiresidue, AOAC, and European procedures.
- Ready to use for QuEChERS extractions-no dilutions necessary
- Support for GC and LC with MS, MS/MS, and selective detectors
- Save time with convenient mixes or make your own blend using our full line of single-component solutions