Order Entry
Germany
ContactUsLinkComponent
Ammonia solution 28.0 - 30.0% (NH₃ basis) in water ACS, Fluka™
Irritant
Corrosive
Ammonia solution 28.0 - 30.0% (NH₃ basis) in water ACS, Fluka™
Certificates
Irritant
Corrosive
Ammonia solution 28.0 - 30.0% (NH₃ basis) in water ACS, Fluka™
Certificates

Specification Test Results

  • Appearance:
    complying
  • Assay (acidimetric):
    28.0 - 30.0 %
  • Residue on ignition:
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.00002 %
  • Heavy metals (as Pb):
    Max. 0.00005 %
  • KMnO₄ reducing matter:
    complying
  • CO₂:
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.00005 %
  • Nitrate (NO₃):
    Max. 0.0002 %
  • Sulfate (SO₄):
    Max. 0.0002 %
  • Phosphate (PO₄):
    Max. 0.0002 %

Specification Test Results

Related Information