Order Entry
Germany
ContactUsLinkComponent
Ammonia solution ≥25% (NH₃ basis) in water, Puriss. p.a. plus, Fluka™
Irritant
Corrosive
Ammonia solution ≥25% (NH₃ basis) in water, Puriss. p.a. plus, Fluka™
Catalog # 17093-1L
CAS Number:  
Irritant
Corrosive
Ammonia solution ≥25% (NH₃ basis) in water, Puriss. p.a. plus, Fluka™
Catalog # 17093-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 25 %
  • Non-volatile matter:
    Max. 10 ppm
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.01 ppm
  • Arsenic, antimony (as As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.01 ppm
  • Boron (B):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.01 ppm
  • Calcium (Ca):
    Max. 0.01 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Cesium (Cs):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.01 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Germanium (Ge):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.01 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.01 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.01 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.01 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.01 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Explosion Limit:
    27 %
  • Chloride (Cl):
    Max. 0.2 ppm
  • Phosphate (PO₄):
    Max. 0.2 ppm
  • Sulfate (SO₄):
    Max. 1 ppm

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:37.7 °C (1.013 hPa)
Melting Pt:-92 °C
Density:0.90 g/cm³ (20 °C)
UN:2672
ADR:8,III
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
Melting Pt:–92 °C