Order Entry
Germany
ContactUsLinkComponent
 
Sodium dihydrogen phosphate monohydrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
 
undefined
Sodium dihydrogen phosphate monohydrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • pH (5 %, 25 °C):
    4.1 - 4.5
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Water insoluble matter:
    Max. 0.01 %
  • Arsenic (As):
    Max. 0.5 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 30 ppm
  • Total nitrogen (as N):
    Max. 10 ppm