Order Entry
Germany
ContactUsLinkComponent
Health
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Certificates
undefined
Health
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Certificates

Specification Test Results

  • Suitability (for KF-titration):
    test passed
  • Water (Karl Fischer):
    Max. 0.02 %
  • Number Of Samples:
    Min. 2
  • Vapor Density:
    1.55 (vs air)
  • Refractive Index:
    n20/D 1.447 (lit.)
  • Vapor Pressure:
    30 mmHg ( 129 °C)
  • Autoignition Temperature:
    932 °F

Specification Test Results

Related Information