


Nickel(II) sulphate hexahydrate 99-102.0%, Puriss. p.a. ACS, Fluka™
Catalog # HONE31483.9050
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Identity (IR):complying
- Appearance:complying
- Assay:99 - 102.0 %
- Water insoluble matter:Max. 0.005 %
- pH (5 %, 20°C):4-6
- Calcium (Ca):Max. 0.005 %
- Copper (Cu):Max. 0.002 %
- Cobalt (Co):Max. 0.002 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.005 %
- Manganese (Mn):Max. 0.0005 %
- Sodium (Na):Max. 0.01 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.002 %
- Chloride (Cl):Max. 0.001 %
Specification Test Results
About this item
:
:262.85 g/mol
Merck Index:13,06541
Melting Pt:53 °C
Density:1.95 g/cm3
Storage Temperature:Ambient
EINECS:231-111-4
UN:3288
ADR:6.1,III
:GHS09
:GHS07
:GHS08
CAS Number:10101-97-0
Formula:NiSO₄·6H₂O
MDL Number:MFCD00149813