Order Entry
Germany
ContactUsLinkComponent
Potassium sulphate 99-101%, meets analytical specification of Ph.Eur., Fluka™
Potassium sulphate 99-101%, meets analytical specification of Ph.Eur., Fluka™
Certificates
Potassium sulphate 99-101%, meets analytical specification of Ph.Eur., Fluka™

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (130°C):
    Max. 0.5 %
  • Acidity or alkalinity:
    complying
  • Calcium (Ca):
    Max. 50 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 20 ppm
  • Sodium (Na):
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 20 ppm
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

Specification Test Results

Frequently Bought Together