Order Entry
Germany
ContactUsLinkComponent
Zinc acetate dihydrate ≥99%, Technical Grade
Zinc acetate dihydrate ≥99%, Technical Grade
Catalog # HONE25056.9025
CAS Number:  
Zinc acetate dihydrate ≥99%, Technical Grade
Catalog # HONE25056.9025
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Iron (Fe):
    Max. 0.01 %
  • Lead (Pb):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :219.51 g/mol
Merck Index:12,10256
Melting Pt:237 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961