Ammonium iron(II) sulphate hexahydrate ≥99% (by manganometry), puriss.
Catalog # HONE12304.9050
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (manganometric):100.5 - 102.4 %
- Calcium (Ca):Max. 0.02 %
- Copper (Cu):Max. 0.002 %
- Iron(III):Max. 0.05 %
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.02 %
- Sodium (Na):Max. 0.01 %
- Zinc (Zn):Max. 0.01 %
- Chloride (Cl):Max. 0.005 %
Specification Test Results
About this item
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
:
:392.14 g/mol
Melting Pt:100 °C
Density:1.864 g/cm³ (20 °C)
EINECS:233-151-8
:GHS07
Storage Temperature:Ambient
CAS Number:7783-85-9
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530