Order Entry
Germany
ContactUsLinkComponent
Silver nitrate concentrate for dilution to 1L, 0.01 M AgNO3 (0.01 N) volumetric solution, Fluka™
Silver nitrate concentrate for dilution to 1L, 0.01 M AgNO3 (0.01 N) volumetric solution, Fluka™
Catalog # 38001-1EA
CAS Number:  
undefined
Silver nitrate concentrate for dilution to 1L, 0.01 M AgNO3 (0.01 N) volumetric solution, Fluka™
Catalog # 38001-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
ADR:8,II
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414