
Orthoboric acid ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. buffer substance, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Melting Pt:171 °C
Density:1.510 g/cm³ (20 °C)
:61.83 g/mol
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:99.8 - 100.5 %
- Appearance of the solution:complying
- Solubility in C₂H₅OH:complying
- Insoluble in methanol:Max. 0.005 %
- With methanol-HCl non-volatile matter:Max. 0.05 %
- pH (4 %, 20°C):3.6 - 4.0
- pH (3,3 %, 20 °C):3.8 - 4.8
- Arsenic (As):Max. 0.00005 %
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0001 %
- Magnesium (Mg):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 0.0003 %
- Phosphate (PO₄):Max. 0.0005 %
- Sulfate (SO₄):Max. 5 ppm
- Organic matter:complying