


Sodium nitrite 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E250, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7632-00-0
:
Formula:NaNO₂
MDL Number:MFCD00011118
:69 g/mol
Merck Index:13,08721
Melting Pt:280 °C
Density:2.17 g/cm3 (20 °C)
EINECS:231-555-9
UN:1500
ADR:5.1,III
:GHS09
:GHS06
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Assay (calculated to the dried substance):99 - 100.5 %
- Loss on drying (vacuum):Max. 0.25 %
- Loss on drying (on silicagel, 4h):Max. 0.2 %
- Acidity or alkalinity:complying
- Arsenic (As):Max. 2 ppm
- Calcium (Ca):Max. 0.005 %
- Mercury (Hg):Max. 1 ppm
- Lead (Pb):Max. 2 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 50 ppm
- Sulfate (SO₄):Max. 100 ppm
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together