Order Entry
Germany
ContactUsLinkComponent
 
Potassium iodide 99%, Reagent Grade
Catalog # 207969-500G
CAS Number:  
Potassium iodide 99%, Reagent Grade
Catalog # 207969-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    98.5 - 101.5 %
  • Vapor Pressure:
    1 mmHg ( 745 °C)

Specification Test Results

About this item

 :
 :166 g/mol
Merck Index:13,07727
Boiling Pt:1.323 °C (1.013 hPa)
Melting Pt:681 °C
Density:3.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-659-4
 :GHS08
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405